missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MOCS2 Protein
A cDNA sequence encoding the MOCS2 was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 1735.00 CHF
Spezifikation
Gen-ID (Entrez) | 4338 |
---|---|
Name | MOCS2 Protein |
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gensymbol | MOCS2 |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15921319
|
enQuireBio™
QP12715-2UG |
2 μg |
131.00 CHF
2 Mikrogramm |
Verfügbar ab: 21-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15911319
|
enQuireBio™
QP12715-10UG |
10 μg |
203.00 CHF
10 Mikrogramm |
Verfügbar ab: 21-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15901319
|
enQuireBio™
QP12715-100UG |
100 μg |
1735.00 CHF
100 Mikrogramm |
Verfügbar ab: 21-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
4338 | |
Research Use Only | |
MOCS2 | |
Human | |
His | |
The MOCS2 solution (0.25 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 50% glycerol. |
MOCS2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMSSL EISSSCFSLE TKLPLSPPLV EDSAFEPSRK DMDEVEEKSK DVINFTAEKL SVDEVSQLVI SPLCGAISLF VGTTRNNFEG KKVISLEYEA YLPMAENEVR KICSDIRQKW PVKHIAVFHR LGLVPVSEAS IIIAVSSAHR AASLEAVSYA IDTLKAKVPI WKKEIYEESS TWKGNKECFW ASNS | |
Greater than 90% as determined by SDS-PAGE. |