missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MMAB Protein

A cDNA sequence encoding the MMAB was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP12702-1mg

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15911279

  • 3735.00 CHF / 1 Milligramm


Verfügbar ab
26-03-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

MMAB Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
MMAB 1 mg/ml protein solution contains 20mM Tris pH 7.5 and 10% glycerol.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MQSRGPQGVE DGDRPQPSSK TPRIPKIYTK TGDKGFSSTF TGERRPKDDQ VFEAVGTTDE LSSAIGFALE LVTEKGHTFA EELQKIQCTL QDVGSALATP CSSAREAHLK YTTFKAGPIL ELEQWIDKYT SQLPPLTAFI LPSGGKISSA LHFCRAVCRR AERRVVPLVQ MGETDANVAK FLNRLSDYLF TLARYAAMKE GNQEKIYKKN DPSAESEGL
Videos
Dokumentation