missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MBD3 Protein
A cDNA sequence encoding the MBD3 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12654-10ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
MBD3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMERKRWE CPALPQGWER EEVPRRSGLS AGHRDVFYYS PSGKKFRSKP QLARYLGGSM DLSTFDFRTG KMLMSKMNKS RQRVRYDSSN QVKGKPDLNT ALPVRQTASI FKQPVTKITN HPSNKVKSDP QKAVDQPRQL FWEKKLSGLN AFDIAEELVK TMDLPKGLQG VGPGCTDETL LSAIASALHT STMPITGQLS AAVEKNPGVW LNTTQPLCKA FMVTDEDIRK QEELVQQVRK RLEEALMADM LAHVEELARD GEAPLDKACA EDDDEEDEEE EEEEPDPDPE MEHV | |
Greater than 85% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The MBD3 solution (0.25 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 1mM DTT, 1mM EDTA and 40% glycerol. |