missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MAVS Protein

A cDNA sequence encoding the MAVS was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP12652-1ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15911119

  • 131.00 CHF / 1 Mikrogramm
Verfügbar ab: 17-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Spezifikation

Spezifikation

MAVS Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMPFAEDK TYKYICRNFS NFCNVDVVEI LPYLPCLTAR DQDRLRATCT LSGNRDTLWH LFNTLQRRPG WVEYFIAALR GCELVDLADE VASVYQSYQP RTSDRPPDPL EPPSLPAERP GPPTPAAAHS IPYNSCREKE PSYPMPVQET QAPESPGENS EQALQTLSPR AIPRNPDGGP LESSSDLAAL SPLTSSGHQE QDTELGSTHT AGATSSLTPS RGPVSPSVSF QPLARSTPRA SRLPGPTGSV VSTGTSFSSS SPGLASAGAA EGKQGAESDQ AEPIICSSGA EAPANSLPSK VPTTLMPVNT VALKVPANPA SVSTVPSKLP TSSKPPGAVP SNALTNPAPS KLPINSTRAG MVPSKVPTSM VLTKVSASTV PTDGSSRNEE TPAAPTPAGA TGGSSAWLDS SSENRGLGSE LSKPGVLASQ VDSPFSGCFE DLAISASTSL GMGPCHGPEE NEYKSEGTFG IHVAENPSIQ LLEGNPGPPA DPDGGPRPQA DRKFQEREVP CHRPSP
Greater than 85% as determined by SDS-PAGE.
1 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The MAVS solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote