missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MAVS Protein
A cDNA sequence encoding the MAVS was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 1735.00 CHF
Spezifikation
Name | MAVS Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Human |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15911119
|
enQuireBio™
QP12652-1UG |
1 μg |
131.00 CHF
1 Mikrogramm |
Verfügbar ab: 17-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15931119
|
enQuireBio™
QP12652-5UG |
5 μg |
203.00 CHF
5 Mikrogramm |
Verfügbar ab: 17-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15921119
|
enQuireBio™
QP12652-50UG |
50 μg |
1735.00 CHF
50 Mikrogramm |
Verfügbar ab: 17-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
MAVS Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The MAVS solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMPFAEDK TYKYICRNFS NFCNVDVVEI LPYLPCLTAR DQDRLRATCT LSGNRDTLWH LFNTLQRRPG WVEYFIAALR GCELVDLADE VASVYQSYQP RTSDRPPDPL EPPSLPAERP GPPTPAAAHS IPYNSCREKE PSYPMPVQET QAPESPGENS EQALQTLSPR AIPRNPDGGP LESSSDLAAL SPLTSSGHQE QDTELGSTHT AGATSSLTPS RGPVSPSVSF QPLARSTPRA SRLPGPTGSV VSTGTSFSSS SPGLASAGAA EGKQGAESDQ AEPIICSSGA EAPANSLPSK VPTTLMPVNT VALKVPANPA SVSTVPSKLP TSSKPPGAVP SNALTNPAPS KLPINSTRAG MVPSKVPTSM VLTKVSASTV PTDGSSRNEE TPAAPTPAGA TGGSSAWLDS SSENRGLGSE LSKPGVLASQ VDSPFSGCFE DLAISASTSL GMGPCHGPEE NEYKSEGTFG IHVAENPSIQ LLEGNPGPPA DPDGGPRPQA DRKFQEREVP CHRPSP | |
Greater than 85% as determined by SDS-PAGE. |