missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MAPK11 Protein
A cDNA sequence encoding the MAPK11 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12640-5ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
MAPK11 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSGPRAG FYRQELNKTV WEVPQRLQGL RPVGSGAYGS VCSAYDARLR QKVAVKKLSR PFQSLIHARR TYRELRLLKH LKHENVIGLL DVFTPATSIE DFSEVYLVTT LMGADLNNIV KCQALSDEHV QFLVYQLLRG LKYIHSAGII HRDLKPSNVA VNEDCELRIL DFGLARQADE EMTGYVATRW YRAPEIMLNW MHYNQTVDIW SVGCIMAELL QGKALFPGSD YIDQLKRIME VVGTPSPEVL AKISSEHART YIQSLPPMPQ KDLSSIFRGA NPLAIDLLGR MLVLDSDQRV SAAEALAHAY FSQYHDPEDE PEAEPYDESV EAKERTLEEW KELTYQEVLS FKPPEPPKPP GSLEIEQ | |
Greater than 90.0% as determined by SDS-PAGE analysis. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MAPK11 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 2mM DTT, 100mM NaCl and 20% glycerol. |