missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MAP2K2 Protein
A cDNA sequence encoding the MAP2K2 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12638-50ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
MAP2K2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMLARRK PVLPALTINP TIAEGPSPTS EGASEANLVD LQKKLEELEL DEQQKKRLEA FLTQKAKVGE LKDDDFERIS ELGAGNGGVV TKVQHRPSGL IMARKLIHLE IKPAIRNQII RELQVLHECN SPYIVGFYGA FYSDGEISIC MEHMDGGSLD QVLKEAKRIP EEILGKVSIA VLRGLAYLRE KHQIMHRDVK PSNILVNSRG EIKLCDFGVS GQLIDSMANS FVGTRSYMAP ERLQGTHYSV QSDIWSMGLS LVELAVGRYP IPPPDAKELE AIFGRPVVDG EEGEPHSISP RPRPPGRPVS GHGMDSRPAM AIFELLDYIV NEPPPKLPNG VFTPDFQEFV NKCLIKNPAE RADLKMLTNH TFIKRSEVEE VDFAGWLCKT LRLNQPGTPT RTAV | |
Greater than 90.0% as determined by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MAP2K2 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl and 10% glycerol. |