missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MAOA Protein
A cDNA sequence encoding the MAOA was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12633-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
MAOA Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMENQEKA SIAGHMFDVV VIGGGISGLS AAKLLTEYGV SVLVLEARDR VGGRTYTIRN EHVDYVDVGG AYVGPTQNRI LRLSKELGIE TYKVNVSERL VQYVKGKTYP FRGAFPPVWN PIAYLDYNNL WRTIDNMGKE IPTDAPWEAQ HADKWDKMTM KELIDKICWT KTARRFAYLF VNINVTSEPH EVSALWFLWY VKQCGGTTRI FSVTNGGQER KFVGGSGQVS ERIMDLLGDQ VKLNHPVTHV DQSSDNIIIE TLNHEHYECK YVINAIPPTL TAKIHFRPEL PAERNQLIQR LPMGAVIKCM MYYKEAFWKK KDYCGCMIIE DEDAPISITL DDTKPDGSLP AIMGFILARK ADRLAKLHKE IRKKKICELY AKVLGSQEAL HPVHYEEKNW CEEQYSGGCY TAYFPPGIMT QYGRVIRQPV GRIFFAGTET ATKWSGYMEG AVEAGERAAR EVLNGLGKVT EKDIWVQEPE SKDVPAVEIT HTFWERNLPS | |
Greater than 80% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MAOA protein solution (0.5 mg/ml) containing 20mM Tris-HCl (pH 8.0) and 10% glycerol. |