missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MAGOHB Protein
A cDNA sequence encoding the MAGOHB was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12629-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
MAGOHB Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAVASDF YLRYYVGHKG KFGHEFLEFE FRPDGKLRYA NNSNYKNDVM IRKEAYVHKS VMEELKRIID DSEITKEDDA LWPPPDRVGR QELEIVIGDE HISFTTSKIG SLIDVNQSKD PEGLRVFYYL VQDLKCLVFS LIGLHFKIKP I | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MAGOHB protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer, (pH 8.0), 1mM DTT, 30% glycerol and 0.15M NaCl. |