missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human M6PR Protein
A cDNA sequence encoding the M6PR was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12616-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
M6PR Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSTEEKTCD LVGEKGKESE KELALVKRLK PLFNKSFEST VGQGSDTYIY IFRVCREAGN HTSGAGLVQI NKSNGKETVV GRLNETHIFN GSNWIMLIYK GGDEYDNHCG KEQRRAVVMI SCNRHTLADN FNPVSEERGK VQDCFYLFEM DSSLACSPEI SH | |
Greater than 85.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The M6PR solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |