missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human M-CSF / CSF-1 Protein
A DNA sequence encoding the Human M-CSF / CSF-1, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10351-ec-250ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_000748.3 | |
M-CSF / CSF-1 Protein | |
Research Use Only | |
CSF1 | |
The ED50 as determined by a cell proliferation assay using murine M-NFS-60 cells is less than 1 ng/ml, corresponding to a specific activity of > 1.0 M 106 IU/mg. | |
Human | |
Untagged | |
0. 2um filtered PBS, pH7.4, lyophilized |
1435 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
EEVSEYCSHM IGSGHLQSLQ RLIDSQMETS CQITFEFVDQ EQLKDPVCYL KKAFLLVQDI MEDTMRFRDN TPNAIAIVQL QELSLRLKSC FTKDYEEHDK ACVRTFYETP LQLLEKVKNV FNETKNLLDK DWNIFSKNCN NSFAECSSQG HERQSEGS | |
Greater than 95% as determined by SDS-PAGE and HPLC. |