missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LYPLAL1 Protein

A cDNA sequence encoding the LYPLAL1 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP12607-5ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15910979

  • 125.00 CHF / 5 Mikrogramm


Verfügbar ab
02-11-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

127018
Greater than 95% as determined by SDS-PAGE.
Research Use Only
LYPLAL1
Human
His
The LYPLAL1 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl and 10% glycerol.
LYPLAL1 Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMAAASG SVLQRCIVSP AGRHSASLIF LHGSGDSGQG LRMWIKQVLN QDLTFQHIKI IYPTAPPRSY TPMKGGISNV WFDRFKITND CPEHLESIDV MCQVLTDLID EEVKSGIKKN RILIGGFSMG GCMAMHLAYR NHQDVAGVFA LSSFLNKASA VYQALQKSNG VLPELFQCHG TADELVLHSW AEETNSMLKS LGVTTKFHSF PNVYHELSKT ELDILKLWIL TKLPGEMEKQ K
Videos
Dokumentation