missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LY9 Protein
A cDNA sequence encoding the LY9 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12603-1ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
LY9 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
ADLKDSAPTV VSGILGGSVT LPLNISVDTE IENVIWIGPK NALAFARPKE NVTIMVKSYL GRLDITKWSY SLCISNLTLN DAGSYKAQIN QRNFEVTTEE EFTLFVYEQL QEPQVTMKSV KVSENFSCNI TLMCSVKGAE KSVLYSWTPR EPHASESNGG SILTVSRTPC DPDLPYICTA QNPVSQRSSL PVHVGQFCTD PGASRGGTTG ETVVGVLGEP VTLPLALPAC RDTEKVVWLF NTSIISKERE EAATADPLIK SRDPYKNRVW VSSQDCSLKI SQLKIEDAGP YHAYVCSEAS SVTSMTHVTL LIYRRLRKPK ITWSLRHSED GICRISLTCS VEDGGNTVMY TWTPLQKEAV VSQGESHLNV SWRSSENHPN LTCTASNPVS RSSHQFLSEN ICSGPERNTK HHHHHH | |
Greater than 90.0% as determined by SDS-PAGE. |
1 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
LY9 protein solution (0.25 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |