missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LHRH Protein
A cDNA sequence encoding the LHRH was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12569-10mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
2796 | |
10 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
Pyr-His-Trp-Ser-Tyr-Gly-Leu-Arg-Pro-Gly-NH2 | |
Greater than 98.0% as determined by RP-HPLC. |
LHRH Protein | |
Research Use Only | |
GNRH1 | |
Human | |
Untagged | |
The LHRH was lyophilized from a concentrated (1 mg/ml) solution with no additives. |