missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Leptin tA Protein
A cDNA sequence encoding the Leptin tA was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10761-50ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
Leptin tA Protein | |
Research Use Only | |
Leptin triple antagonist is capable of inhibiting leptin-induced proliferation of BAF/3 cells stably transfected with the long form of human leptin receptor. It also inhibits various leptin effects in several in vitro bioassays. | |
Human | |
Untagged | |
Greater than 98.0% as determined by:(a) Gel filtration analysis. (b) Analysis by SDS-PAGE. |
50 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.0045mM NaHCO3. |