missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LAMP2 Protein

A cDNA sequence encoding the LAMP2 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP12530-2ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15910729

  • 125.00 CHF / 2 Mikrogramm


Verfügbar ab
02-11-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

LAMP2 Protein
Research Use Only
Recombinant Protein
Insect
MVCFRLFPVP GSGLVLVCLV LGAVRSYALE LNLTDSENAT CLYAKWQMNF TVRYETTNKT YKTVTISDHG TVTYNGSICG DDQNGPKIAV QFGPGFSWIA NFTKAASTYS IDSVSFSYNT GDNTTFPDAE DKGILTVDEL LAIRIPLNDL FRCNSLSTLE KNDVVQHYWD VLVQAFVQNG TVSTNEFLCD KDKTSTVAPT IHTTVPSPTT TPTPKEKPEA GTYSVNNGND TCLLATMGLQ LNITQDKVAS VININPNTTH STGSCRSHTA LLRLNSSTIK YLDFVFAVKN ENRFYLKEVN ISMYLVNGSV FSIANNNLSY WDAPLGSSYM CNKEQTVSVS GAFQINTFDL RVQPFNVTQG KYSTAQDCSA DDDNFLVPIA VGAALAGVLI LVLLAYFIGL KHHHAGYEQF
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
LAMP2 (0.94 mg/ml) is supplied in 16mM HEPES buffer pH 8.0, 130mM NaCl and 20% Glycerol.
Videos
Dokumentation