missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LAGE3 Protein
A cDNA sequence encoding the LAGE3 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12524-100ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
8270 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMRDADAD AGGGADGGDG RGGHSCRGGV DTAAAPAGGA PPAHAPGPGR DAASAARGSR MRPHIFTLSV PFPTPLEAEI AHGSLAPDAE PHQRVVGKDL TVSGRILVVR WKAEDCRLLR ISVINFLDQL SLVVRTMQRF GPPVSR | |
Greater than 85% as determined by SDS-PAGE. |
LAGE3 Protein | |
Research Use Only | |
LAGE3 | |
Human | |
His | |
The LAGE3 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 50% glycerol and 2mM DTT. |