missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KLRD1 Protein
A cDNA sequence encoding the KLRD1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12511-50ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
KLRD1 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
ADPKNSFTKL SIEPAFTPGP NIELQKDSDC CSCQEKWVGY RCNCYFISSE QKTWNESRHL CASQKSSLLQ LQNTDELDFM SSSQQFYWIG LSYSEEHTAW LWENGSALSQ YLFPSFETFN TKNCIAYNPN GNALDESCED KNRYICKQQL IHHHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
KLRD1 protein solution (0.5 mg/ml) contains Phosphate buffered saline (pH 7.4) and 10% glycerol. |