missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KLRB1 Protein
A cDNA sequence encoding the KLRB1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12508-HIS-20ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
KLRB1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMQKSSIE KCSVDIQQSR NKTTERPGLL NCPIYWQQLR EKCLLFSHTV NPWNNSLADC STKESSLLLI RDKDELIHTQ NLIRDKAILF WIGLNFSLSE KNWKWINGSF LNSNDLEIRG DAKENSCISI SQTSVYSEYC STEIRWICQK ELTPVRNKVY PDS | |
Greater than 85% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The KLRB1 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 4M urea and 10% glycerol. |