missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KIR2DL1 Protein
A cDNA sequence encoding the KIR2DL1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12487-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
KIR2DL1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MEGVHRKPSL LAHPGRLVKS EETVILQCWS DVMFEHFLLH REGMFNDTLR LIGEHHDGVS KANFSISRMT QDLAGTYRCY GSVTHSPYQV SAPSDPLDIV IIGLYEKPSL SAQLGPTVLA GENVTLSCSS RSSYDMYHLS REGEAHERRL PAGPKVNGTF QADFPLGPAT HGGTYRCFGS FHDSPYEWSK SSDPLLVSVT GN | |
Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The protein (1 mg/ml) contains 25mM Tris- HCl (pH 7.5). |