missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IYD Protein

A cDNA sequence encoding the IYD was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP12467-1mg

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15910519

  • 3735.00 CHF / 1 Milligramm


Verfügbar ab
23-10-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

IYD Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
IYD protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA and 10% glycerol.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSDRSMEKK KGEPRTRAEA RPWVDEDLKD SSDLHQAEED ADEWQESEEN VEHIPFSHNH YPEKEMVKRS QEFYELLNKR RSVRFISNEQ VPMEVIDNVI RTAGTAPSGA HTEPWTFVVV KDPDVKHKIR KIIEEEEEIN YMKRMGHRWV TDLKKLRTNW IKEYLDTAPI LILIFKQVHG FAANGKKKVH YYNE
Videos
Dokumentation