missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ITGB1BP3 Protein
A cDNA sequence encoding the ITGB1BP3 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12463-2ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
27231 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMKLIVGI GGMTNGGKTT LTNSLLRALP NCCVIHQDDF FKPQDQIAVG EDGFKQWDVL ESLDMEAMLD TVQAWLSSPQ KFARAHGVSV QPEASDTHIL LLEGFLLYSY KPLVDLYSRR YFLTVPYEEC KWRRSTRNYT VPDPPGLFDG HVWPMYQKYR QEMEANGVEV VYLDGMKSRE ELFREVLEDI QNSLLNRSQE SAPSPARPAR TQGPGRGCGH RTARPAASQQ DSM | |
Greater than 90% as determined by SDS-PAGE. |
ITGB1BP3 Protein | |
Research Use Only | |
NMRK2 | |
Human | |
His | |
The ITGB1BP3 solution (0.25 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 1mM DTT and 40% glycerol. |