missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Involucrin Protein
A cDNA sequence encoding the Involucrin was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12453-10ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
3713 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MSQQHTLPVT LSPALSQELL KTVPPPVNTH QEQMKQPTPL PPPCQKVPVE LPVEVPSKQE EKHMTAVKGL PEQECEQQQK EPQEQELQQQ HWEQHEEYQK AENPEQQLKQ EKTQRDQQLN KQLEEEKKLL DQQLDQELVK RDEQLGMKKE QLLELPEQQE GHLKHLEQQE GQLKHPEQQE GQLELPEQQE GQLELPEQQE GQLELPEQQE GQLELPEQQE GQLELPEQQE GQLELPQQQE GQLELSEQQE GQLELSEQQE GQLKHLEHQE GQLEVPEEQM GQLKYLEQQE GQLKHLDQQE KQPELPEQQM GQLKHLEQQE GQPKHLEQQE GQLEQLEEQE GQLKHLEQQE GQLEHLEHQE GQLGLPEQQV LQLKQLEKQQ GQPKHLEEEE GQLKHLVQQE GQLKHLVQQE GQLEQQERQV EHLEQQVGQL KHLEEQEGQL KHLEQQQGQL EVPEQQVGQP KNLEQEEKQL ELPEQQEGQV KHLEKQEAQL ELPEQQVGQP KHLEQQEKHL EHPEQQDGQL KHLEQQEGQL KDLEQQKGQL EQPVFAPAPG QVQDIQPALP TKGEVLLPVE HQQQKQEVQW PPKHK |
Involucrin Protein | |
Research Use Only | |
IVL | |
Human | |
GST | |
Involucrin at 100microg/ml in 50mM Tris-HCl, pH 7.5, 10mM L-glutathione (reduced). |