missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL36B / IL1F8 Protein
A DNA sequence encoding the Human IL36B / IL1F8, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10339-ec-100ug
Weitere Details : Gewicht : 0.01000kg
Spezifikation
NP_055253.2 | |
IL36B / IL1F8 Protein | |
Research Use Only | |
Il36b | |
The specific activity is determined by its binding ability in a functional ELISA. Immobilized rHuIL-36beta at 1 ug/mL can bind recombinant human IL-1 Rrp2 Fc Chimera with a range of 0.15-5 ug/mL. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
27177 | |
100 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MNPQREAAPK SYAIRDSRQM VWVLSGNSLI AAPLSRSIKP VTLHLIACRD TEFSDKEKGN MVYLGIKGKD LCLFCAEIQG KPTLQLKEKN IMDLYVEKKA QKPFLFFHNK EGSTSVFQSV SYPGWFIATS TTSGQPIFLT KERGITNNTN FYLDSVE | |
Greater than 97% as determined by SDS-PAGE and HPLC. |