missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL35 Protein

A cDNA sequence encoding the IL35 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP12423-10ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15910379

  • 188.00 CHF / 10 Mikrogramm


Verfügbar ab
02-11-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

IL35 Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
Lyophilized from a concentrated solution containing 10mM Na2PO4 pH 7.5 and 30mM NaCl.
Greater than 95.0% as determined by analysis by SDS-PAGE.
Research Use Only
Recombinant Protein
HEK 293
EBI3 subunit:RKGPPAALTL PRVQCRASRY PIAVDCSWTL PPAPNSTSPV SFIATYRLGM AARGHSWPCL QQTPTSTSCT ITDVQLFSMA PYVLNVTAVH PWGSSSSFVP FITEHIIKPD PPEGVRLSPL AERQLQVQWE PPGSWPFPEI FSLKYWIRYK RQGAARFHRV GPIEATSFIL RAVRPRARYY VQVAAQDLTD YGELSDWSLP ATATMSLGK p35 subunit:RNLPVATPDP GMFPCLHHSQ NLLRAVSNML QKARQTLEFY PCTSEEIDHE DITKDKTSTV EACLPLELTK NESCLNSRET SFITNGSCLA SRKTSFMMAL CLSSIYEDLK MYQVEFKTMN AKLLMDPKRQ IFLDQNMLAV IDELMQALNF NSETVPQKSS LEEPDFYKTK IKLCILLHAF RIRAVTIDRV MSYLNAS
Videos
Dokumentation