missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL33 / Interleukin-33 / NF-HEV Protein

A DNA sequence encoding the Human IL33 / Interleukin-33 / NF-HEV Protein, was expressed in the hosts and tags indicated.

Marke:  enQuireBio™ QP10329-ec-10ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15983518

  • 313.00 CHF / 10 Mikrogramm


Verfügbar ab
30-10-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

NP_001186569.1
IL33 / Interleukin-33 / NF-HEV Protein
10 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
SITGISPITE YLASLSTYND QSITFALEDE SYEIYVEDLK KDEKKDKVLL SYYESQHPSN ESGDGVDGKM LMVTLSPTKD FWLHANNKEH SVELHKCEKP LPDQAFFVLH NMHSNCVSFE CKTDPGVFIG VKDNHLALIK VDSSENLCTE NILFKLSET
90865
Greater than 97% as determined by SDS-PAGE and HPLC.
Research Use Only
IL33
The ED50 as determined by a cell proliferation assay using murine D10S cells is less than 0.05 ng/ml, corresponding to a specific activity of > 2.0 M 107 IU/mg.
Human
Untagged
0.2um filtered 20 mM PBS, 150 mM NaCl, 1mM EDTA, pH7.4, lyophilized
Videos
Dokumentation