missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL19 / Interleukin-19 Protein

A DNA sequence encoding the Human IL19 / Interleukin-19, was expressed in the hosts and tags indicated.

Marke:  enQuireBio™ QP10324-ec-10ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15983498

  • 313.00 CHF / 10 Mikrogramm


Verfügbar ab
29-10-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

NP_037503.2
IL19 / Interleukin-19 Protein
10 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
LRRCLISTDM HHIEESFQEI KRAIQAKDTF PNVTILSTLE TLQIIKPLDV CCVTKNLLAF YVDRVFKDHQ EPNPKILRKI SSIANSFLYM QKTLRQCQEQ RQCHCRQEAT NATRVIHDNY DQLEVHAAAI KSLGELDVFL AWINKNHEVM SSA
29949
Greater than 95% as determined by SDS-PAGE and HPLC.
Research Use Only
IL19
The ED50 as determined by a cell proliferation assay using human IL-20Ralpha and human IL-20Rbeta co-transfected murine BaF3 pro-B cells is less than 1.5 ng/ml, corresponding to a specific activity of > 6.7M 105 IU/mg.
Human
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
Dokumentation