missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL19 / Interleukin-19 Protein
A DNA sequence encoding the Human IL19 / Interleukin-19, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10324-ec-250ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_037503.2 | |
IL19 / Interleukin-19 Protein | |
Research Use Only | |
IL19 | |
The ED50 as determined by a cell proliferation assay using human IL-20Ralpha and human IL-20Rbeta co-transfected murine BaF3 pro-B cells is less than 1.5 ng/ml, corresponding to a specific activity of > 6.7M 105 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
29949 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
LRRCLISTDM HHIEESFQEI KRAIQAKDTF PNVTILSTLE TLQIIKPLDV CCVTKNLLAF YVDRVFKDHQ EPNPKILRKI SSIANSFLYM QKTLRQCQEQ RQCHCRQEAT NATRVIHDNY DQLEVHAAAI KSLGELDVFL AWINKNHEVM SSA | |
Greater than 95% as determined by SDS-PAGE and HPLC. |