missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL13 / ALRH Protein
A DNA sequence encoding the Human IL13 / ALRH, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10320-ec-2ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_002179.2 | |
IL13 / ALRH Protein | |
Research Use Only | |
IL13 | |
The ED50 as determined by a cell proliferation assay using human TF-1 cells is less than 0.5 ng/ml, corresponding to a specific activity of > 2.0 M 106 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.2, with 5 % trehalose, lyophilized |
3596 | |
2 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
GPVPPSTALR ELIEELVNIT QNQKAPLCNG SMVWSINLTA GMYCAALESL INVSGCSAIE KTQRMLSGFC PHKVSAGQFS SLHVRDTKIE VAQFVKDLLL HLKKLFREGQ FN | |
Greater than 97% as determined by SDS-PAGE and HPLC. |