missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL10 / Interleukin-10 Protein
A DNA sequence encoding the Human IL10 / Interleukin-10, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10318-ec-2ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_000563.1 | |
IL10 / Interleukin-10 Protein | |
Research Use Only | |
IL10 | |
The ED50 as determined by a cell proliferation assay using murine MC/9-2 cells is less than 1 ng/ml, corresponding to a specific activity of > 1.0 M 106 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
3586 | |
2 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
SPGQGTQSEN SCTHFPGNLP NMLRDLRDAF SRVKTFFQMK DQLDNLLLKE SLLEDFKGYL GCQALSEMIQ FYLEEVMPQA ENQDPDIKAH VNSLGENLKT LRLRLRRCHR FLPCENKSKA VEQVKNAFNK LQEKGIYKAM SEFDIFINYI EAYMTMKIRN | |
Greater than 95% as determined by SDS-PAGE and HPLC. |