missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL 28B Protein
A cDNA sequence encoding the IL 28B was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 7525.00 CHF
Spezifikation
Gen-ID (Entrez) | 282617 |
---|---|
Name | IL 28B Protein |
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gensymbol | IFNL3 |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15984928
|
enQuireBio™
QP10709-2UG |
2 μg |
131.00 CHF
2 Mikrogramm |
Verfügbar ab: 21-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15964928
|
enQuireBio™
QP10709-10UG |
10 μg |
203.00 CHF
10 Mikrogramm |
Verfügbar ab: 21-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15974928
|
enQuireBio™
QP10709-1MG |
1 mg |
7525.00 CHF
1 Milligramm |
Verfügbar ab: 21-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
282617 | |
Research Use Only | |
IFNL3 | |
Recombinant Protein | |
Human (HEK293) | |
MTGDCMPVLV LMAAVLTVTG AVPVARLRGA LPDARGCHIA QFKSLSPQEL QAFKRAKDAL EESLLLKDCK CRSRLFPRTW DLRQLQVRER PVALEAELAL TLKVLEATAD TDPALGDVLD QPLHTLHHIL SQLRACIQPQ PTAGPRTRGR LHHWLHRLQE APKKESPGCL EASVTFNLFR LLTRDLNCVA SGDLCV | |
Greater than 95% as obsereved by SDS-PAGE. |
IL 28B Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The specific activity was determined by the dose dependent protection of the cytopathic effect on A549 cells (human lung adenocarcinoma epithelial cell line) that were challenged with encephalomyocarditis (EMC) virus and is typically 0.1-1ng/ml. | |
Human | |
Untagged | |
The IL28B was lyophilized from 1 mg/ml in 1xPBS. |