missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL-1 beta / IL1B Protein

A DNA sequence encoding the Human IL-1 beta / IL1B, was expressed in the hosts and tags indicated.

Marke:  enQuireBio™ QP10307-ec-100ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15983438

  • 1670.00 CHF / 100 Mikrogramm


Verfügbar ab
26-10-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

NP_000567.1
IL-1 beta / IL1B Protein
100 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
APVRSLNCTL RDSQQKSLVM SGPYELKALH LQGQDMEQQV VFSMSFVQGE ESNDKIPVAL GLKEKNLYLS CVLKDDKPTL QLESVDPKNY PKKKMEKRFV FNKIEINNKL EFESAQFPNW YISTSQAENM PVFLGGTKGG QDITDFTMQF VSS
3553
Greater than 97% as determined by SDS-PAGE and HPLC.
Research Use Only
IL1B
The ED50 as determined by a cell proliferation assay using murine D10S cells is less than 1.0 pg/ml, corresponding to a specific activity of > 1.0 M 109 IU/mg.
Human
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
Dokumentation