missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL-1 beta / IL1B Protein
A DNA sequence encoding the Human IL-1 beta / IL1B, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10307-ec-10ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_000567.1 | |
IL-1 beta / IL1B Protein | |
Research Use Only | |
IL1B | |
The ED50 as determined by a cell proliferation assay using murine D10S cells is less than 1.0 pg/ml, corresponding to a specific activity of > 1.0 M 109 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
3553 | |
10 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
APVRSLNCTL RDSQQKSLVM SGPYELKALH LQGQDMEQQV VFSMSFVQGE ESNDKIPVAL GLKEKNLYLS CVLKDDKPTL QLESVDPKNY PKKKMEKRFV FNKIEINNKL EFESAQFPNW YISTSQAENM PVFLGGTKGG QDITDFTMQF VSS | |
Greater than 97% as determined by SDS-PAGE and HPLC. |