missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IGSF8 Protein
A cDNA sequence encoding the IGSF8 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12389-50ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
IGSF8 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
ADLREVLVPE GPLYRVAGTA VSISCNVTGY EGPAQQNFEW FLYRPEAPDT ALGIVSTKDT QFSYAVFKSR VVAGEVQVQR LQGDAVVLKI ARLQAQDAGI YECHTPSTDT RYLGSYSGKV ELRVLPDVLQ VSAAPPGPRG RQAPTSPPRM TVHEGQELAL GCLARTSTQK HTHLAVSFGR SVPEAPVGRS TLQEVVGIRS DLAVEAGAPY AERLAAGELR LGKEGTDRYR MVVGGAQAGD AGTYHCTAAE WIQDPDGSWA QIAEKRAVLA HVDVQTLSSQ LAVTVGPGER RIGPGEPLEL LCNVSGALPP AGRHAAYSVG WEMAPAGAPG PGRLVAQLDT EGVGSLGPGY EGRHIAMEKV ASRTYRLRLE AARPGDAGTY RCLAKAYVRG SGTRLREAAS ARSRPLPVHV REEGVVLEAV AWLAGGTVYR GETASLLCNI SVRGGPPGLR LAASWWVERP EDGELSSVPA QLVGGVGQDG VAELGVRPGG GPVSVELVGP RSHRLRLHSL GPEDEGVYHC APSAWVQHAD YSWYQAGSAR SGPVTVYPYM HALDTHHHHH H | |
Greater than 95.0% as determined by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
IGSF8 protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 30% glycerol. |