missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IGFBP 6 Protein
A cDNA sequence encoding the IGFBP 6 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12381-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
3489 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MTPHRLLPPL LLLLALLLAA SPGGALARCP GCGQGVQAGC PGGCVEEEDG GSPAEGCAEA EGCLRREGQE CGVYTPNCAP GLQCHPPKDD EAPLRALLLG RGRCLPARAP AVAEENPKES KPQAGTARPQ DVNRRDQQRN PGTSTTPSQP NSAGVQDTEM GPCRRHLDSV LQQLQTEVYR GAQTLYVPNC DHRGFYRKRQ CRSSQGQRRG PCWCVDRMGK SLPGSPDGNG SSSCPTGSSG | |
Greater than 80.0% as determined by SDS-PAGE. |
IGFBP 6 Protein | |
Research Use Only | |
IGFBP6 | |
Human | |
His | |
IBP-6 was lyophilized from a concentrated (1 mg/ml) solution containing 1xPBS, 0.1% SDS and 1mM DTT. |