Learn More
enQuireBio™ Recombinant Human IFNB1 / IFN-beta / Interferon beta Protein
A DNA sequence encoding the Human IFNB1 / IFN-beta / Interferon beta, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10408-ec-10ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_002167.1 | |
IFNB1 / IFN-beta / Interferon beta Protein | |
Research Use Only | |
IFNB1 | |
The specific activity determined by an anti-viral assay is no less than 3.0 M 107 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, containing 2 % HSA and 3 % mannitol, lyophilized |
3456 | |
10 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MSYNLLGFLQ RSSNFQCQKL LWQLNGRLEY CLKDRMNFDI PEEIKQLQQF QKEDAALTIY EMLQNIFAIF RQDSSSTGWN ETIVENLLAN VYHQINHLKT VLEEKLEKED FTRGKLMSSL HLKRYYGRIL HYLKAKEYSH CAWTIVRVEI LRNFYFINRL TGYLRN | |
Greater than 97% as determined by SDS-PAGE and HPLC. |