missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IFN-alpha / IFNA1 / IFN Protein
A DNA sequence encoding the Human IFN-alpha / IFNA1 / IFN, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10404-ec-10ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_008831.3 | |
IFN-alpha / IFNA1 / IFN Protein | |
Research Use Only | |
IFNA1 | |
The specific activity determined by an anti-viral assay is no less than 1.0 M 108 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, containing 4 % mannitol and 1 % HSA, lyophilized |
3439 | |
10 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MCDLPETHSL DNRRTLMLLA QMSRISPSSC LMDRHDFGFP QEEFDGNQFQ KAPAISVLHE LIQQIFNLFT TKDSSAAWDE DLLDKFCTEL YQQLNDLEAC VMQEERVGET PLMNVDSILA VKKYFRRITL YLTEKKYSPC AWEVVRAEIM RSLSLSTNLQ ERLRRKE | |
Greater than 96% as determined by SDS-PAGE and HPLC. |