missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IFI30 Protein

A cDNA sequence encoding the IFI30 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP12372-1mg

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15910209

  • 7190.00 CHF / 1 Milligramm


Verfügbar ab
19-03-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

10437
Greater than 90% as determined by SDS-PAGE.
Research Use Only
IFI30
Human
His
The IFI30 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 1mM DTT and 10% glycerol.
IFI30 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMNAPLVN VTLYYEALCG GCRAFLIREL FPTWLLVMEI LNVTLVPYGN AQEQNVSGRW EFKCQHGEEE CKFNKVEACV LDELDMELAF LTIVCMEEFE DMERSLPLCL QLYAPGLSPD TIMECAMGDR GMQLMHANAQ RTDALQPPHE YVPWVTVNGK PLEDQTQLLT LVCQLYQGK
Videos
Dokumentation