missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HTF Protein
A cDNA sequence encoding the HTF was constructed and used to recombinantly synthesize the protein.
372.00 CHF - 2315.00 CHF
Spezifikation
Gen-ID (Entrez) | 7018 |
---|---|
Name | HTF Protein |
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gensymbol | TF |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15910149
|
enQuireBio™
QP12352-500MG |
500 mg |
372.00 CHF
500 Milligramm |
Verfügbar ab: 27-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15900149
|
enQuireBio™
QP12352-1GRAM |
1 g |
580.00 CHF
1 Gramm |
Verfügbar ab: 27-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15920149
|
enQuireBio™
QP12352-5GRAM |
5 g |
2315.00 CHF
5 Gramm |
|
|||||
Spezifikation
7018 | |
Research Use Only | |
TF | |
Human | |
Untagged | |
The protein (10 mg/ml) was lyophilized from 20mM NH4HC03 solution. May contain traces of buffer salts. |
HTF Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Human | |
MRLAVGALLV CAVLGLCLAV PDKTVRWCAV SEHEATKCQS FRDHMKSVIP SDGPSVACVK KASYLDCIRA IAANEADAVT LDAGLVYDAY LAPNNLKPVV AEFYGSKEDP QTFYYAVAVV KKDSGFQMNQ LRGKKSCHTG LGRSAGWNIP IGLLYCDLPE PRKPLEKAVA NFFSGSCAPC ADGTDFPQLC QLCPGCGCST LNQYFGYSGA FKCLKDGAGD VAFVKHSTIF ENLANKADRD QYELLCLDNT RKPVDEYKDC HLAQVPSHTV VARSMGGKED LIWELLNQAQ EHFGKDKSKE FQLFSSPHGK DLLFKDSAHG FLKVPPRMDA KMYLGYEYVT AIRNLREGTC PEAPTDECKP VKWCALSHHE RLKCDEWSVN SVGKIECVSA ETTEDCIAKI MNGEADAMSL DGGFVYIAGK CGLVPVLAEN YNKSDNCEDT PEAGYFAIAV VKKSASDLTW DNLKGKKSCH TAVGRTAGWN IPMGLLYNKI NHCRFDEFFS EGCAPGSKKD SSLCKLCMGS GLNLCEPNNK EGYYGYTGAF RCLVEKGDVA FVKHQTVPQN TGGKNPDPWA KNLNEKDYEL LCLDGTRKPV EEYANCHLAR APNHAVVTRK DKEACVHKIL RQQQHLFGSN VTDCSGNFCL FRSETKDLLF RDDTVCLAKL HDRNTYEKYL GEEYVKAVGN LRKCSTSSLL EACTFRRP | |
Greater than 98.0% as determined by coomassie blue stained SDS-PAGE and Cellulose Acetate electrophoresis. |