missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HSPA1B Protein

A cDNA sequence encoding the HSPA1B was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP12328-1mg

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15910079

  • 7190.00 CHF / 1 Milligramm


Verfügbar ab
06-05-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

HSPA1B Protein
Research Use Only
Recombinant Protein
E. coli
MKHHHHHHAS AKAAAIGIDL GTTYSCVGVF QHGKVEIIAN DQGNRTTPSY VAFTDTERLI GDAAKNQVAL NPQNTVFDAK RLIGRKFGDP VVQSDMKHWP FQVINDGDKP KVQVSYKGET KAFYPEEISS MVLTKMKEIA EAYLGYPVTN AVITVPAYFN DSQRQATKDA GVIAGLNVLR IINEPTAAAI AYGLDRTGKG ERNVLIFDLG GGTFDVSILT IDDGIFEVKA TAGDTHLGGEDFDNRLVNHF VEEFKRKHKK DISQNKRAVR RLRTACERAK RTLSSSTQAS LEIDSLFEGI DFYTSITRAR FEELCSDLFR STLEPVEKAL RDAKLDKAQI HDLVLVGGST RIPKVQKLLQ DFFNGRDLNK SINPDEAVAY GAAVQAAILM GDKSENVQDL LLLDVAPLSL GLETAGGVMT ALIKRNSTIP TKQTQIFTTY SDNQPGVLIQ VYEGERAMTK DNNLLGRFEL SGIPPAPRGVPQIEVTFDID ANGILNVTAT DKSTGKANKI TITNDKGRLS KEEIERMVQE AEKYKAEDEV QRERVSAKNA LESYAFNMKS AVEDEGLKGK ISEADKKKVL DKCQEVISWL DANTLAEKDE FEHKRKELEQ VCNPIISGLY QGAGGPGPGG FGAQGPKGGS GSGPTIEEVD
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The protein was filtered (0.4microm) and lyophilized from a 0.5 mg/ml solution containing 50mM phosphate buffer, 75mM NaCl, pH 7.4.
Videos
Dokumentation