missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HSPA1B Protein
A cDNA sequence encoding the HSPA1B was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 7525.00 CHF
Spezifikation
Name | HSPA1B Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Human |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15920079
|
enQuireBio™
QP12328-2UG |
2 μg |
131.00 CHF
2 Mikrogramm |
Verfügbar ab: 24-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15900079
|
enQuireBio™
QP12328-10UG |
10 μg |
203.00 CHF
10 Mikrogramm |
Verfügbar ab: 24-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15910079
|
enQuireBio™
QP12328-1MG |
1 mg |
7525.00 CHF
1 Milligramm |
Verfügbar ab: 24-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
HSPA1B Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The protein was filtered (0.4microm) and lyophilized from a 0.5 mg/ml solution containing 50mM phosphate buffer, 75mM NaCl, pH 7.4. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHAS AKAAAIGIDL GTTYSCVGVF QHGKVEIIAN DQGNRTTPSY VAFTDTERLI GDAAKNQVAL NPQNTVFDAK RLIGRKFGDP VVQSDMKHWP FQVINDGDKP KVQVSYKGET KAFYPEEISS MVLTKMKEIA EAYLGYPVTN AVITVPAYFN DSQRQATKDA GVIAGLNVLR IINEPTAAAI AYGLDRTGKG ERNVLIFDLG GGTFDVSILT IDDGIFEVKA TAGDTHLGGEDFDNRLVNHF VEEFKRKHKK DISQNKRAVR RLRTACERAK RTLSSSTQAS LEIDSLFEGI DFYTSITRAR FEELCSDLFR STLEPVEKAL RDAKLDKAQI HDLVLVGGST RIPKVQKLLQ DFFNGRDLNK SINPDEAVAY GAAVQAAILM GDKSENVQDL LLLDVAPLSL GLETAGGVMT ALIKRNSTIP TKQTQIFTTY SDNQPGVLIQ VYEGERAMTK DNNLLGRFEL SGIPPAPRGVPQIEVTFDID ANGILNVTAT DKSTGKANKI TITNDKGRLS KEEIERMVQE AEKYKAEDEV QRERVSAKNA LESYAFNMKS AVEDEGLKGK ISEADKKKVL DKCQEVISWL DANTLAEKDE FEHKRKELEQ VCNPIISGLY QGAGGPGPGG FGAQGPKGGS GSGPTIEEVD |