missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HMGN3 Protein
A cDNA sequence encoding the HMGN3 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12286-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
HMGN3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMPKRKSP ENTEGKDGSK VTKQEPTRRS ARLSAKPAPP KPEPKPRKTS AKKEPGAKIS RGAKGKKEEK QEAGKEGTEN | |
Greater than 95% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
HMGN3 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 7.0) and 10% glycerol. |