missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HGF Protein
A cDNA sequence encoding the HGF was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 6225.00 CHF
Spezifikation
Name | HGF Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biologische Aktivität | The specific activity was determined by the dose-dependent stimulation of the proliferation of the monkey epithelial cell line 4MBr-5 and is typically 10-50ng/ml. |
Produkttyp | Recombinant Protein |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15974578
|
enQuireBio™
QP10665-INSECT-2UG |
2 μg |
131.00 CHF
2 Mikrogramm |
Verfügbar ab: 13-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15954578
|
enQuireBio™
QP10665-INSECT-10UG |
10 μg |
203.00 CHF
10 Mikrogramm |
Verfügbar ab: 13-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15964578
|
enQuireBio™
QP10665-INSECT-1MG |
1 mg |
6225.00 CHF
1 Milligramm |
Verfügbar ab: 13-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
HGF Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Baculovirus | |
32-QRKRRNTIHE FKKSAKTTLI KIDPALKIKT KKVNTADQCA NRCTRNKGLP FTCKAFVFDK ARKQCLWFPF NSMSSGVKKE FGHEFDLYEN KDYIRNCIIG KGRSYKGTVS ITKSGIKCQP WSSMIPHEHS FLPSSYRGKD LQENYCRNPR GEEGGPWCFT SNPEVRYEVC DIPQCSEVEC MTCNGESYRG LMDHTESGKI CQRWDHQTPH RHKFLPERYP DKGFDDNYCR NPDGQPRPWC YTLDPHTRWE YCAIKTCADN TMNDTDVPLE TTECIQGQGE GYRGTVNTIW NGIPCQRWDS QYPHEHDMTP ENFKCKDLRE NYCRNPDGSE SPWCFTTDPN IRVGYCSQIP NCDMSHGQDC YRGNGKNYMG NLSQTRSGLT CSMWDKNMED LHRHIFWEPD ASKLNENYCR NPDDDAHGPW CYTGNPLIPW DYCPISRCEG DTTPTIVNLD HPVISCAKTK QLRVVNGIPT RTNIGWMVSL RYRNKHICGG SLIKESWVLT ARQCFPSRDL KDYEAWLGIH DVHGRGDEKC KQVLNVSQLV YGPEGSDLVL MKLARPAVLD DFVSTIDLPN YGCTIPEKTS CSVYGWGYTG LINYDGLLRV AHLYIMGNEK CSQHHRGKVT LNESEICAGA EKIGSGPCEG DYGGPLVCEQ HKMRMVLGVI VPGRGCAIPN RPGIFVRVAY YAKWIHKIIL TYKVPQS-728 | |
Greater than 95% as obsereved by SDS-PAGE. |
Research Use Only | |
The specific activity was determined by the dose-dependent stimulation of the proliferation of the monkey epithelial cell line 4MBr-5 and is typically 10-50ng/ml. | |
Human | |
Untagged | |
The HGF was lyophilized from 1 mg/ml in 1xPBS. |