missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Heterogeneous nuclear ribonucleoprotein 1
A DNA sequence encoding the Homo sapiens (Human) Heterogeneous nuclear ribonucleoprotein K, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP8415-ec-200ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_001305115.1 | |
Heterogeneous nuclear ribonucleoprotein 1 | |
Research Use Only | |
HNRPK | |
Recombinant Protein | |
E. coli | |
TEQPEETFPN TETNGEFGKR PAEDMEEEQA FKRSRNTDEM VELRILLQSK NAGAVIGKGG KNIKALRTDY NASVSVPDSS GPERILSISA DIETIGEILK KIIPTLEEGL QLPSPTATSQ LPLESDAVEC LNYQHYKGSD FDCELRLLIH QSLAGGIIGV KGAKIKELRE NTQTTIKLFQ ECCPHSTDRV VLIGGKPDRV VECIKIILDL ISESPIKGRA QPYDPNFYDE TYDYGGFTMM FDDRRGRPVG FPMRGRGGFD RMPPGRGGRP MPPSRRDYDD MSPRRGPPPP PPGRGGRGGS RARNLPLPPP PPPRGGDLMA YDRRGRPGDR YDGMVGFSAD ETWDSAIDTW SPSEWQMAYE PQGGSGYDYS YAGGRGSYGD LGGPIITTQV TIPKDLAGSI IGKGGQRIKQ IRHESGASIK IDEPLEGSED RIITITGTQD QIQNAQYLLQ NSVKQYS | |
Greater than 90% as determined by SDS-PAGE. |
3190 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |