missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HDHD2 Protein
A cDNA sequence encoding the HDHD2 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12209-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
84064 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAACRALKAV LVDLSGTLHI EDAAVPGAQE ALKRLRGASV IIRFVTNTTK ESKQDLLERL RKLEFDISED EIFTSLTAAR SLLERKQVRP MLLVDDRALP DFKGIQTSDP NAVVMGLAPE HFHYQILNQA FRLLLDGAPL IAIHKARYYK RKDGLALGPG PFVTALEYAT DTKATVVGKP EKTFFLEALR GTGCEPEEAV MIGDDCRDDV GGAQDVGMLG ILVKTGKYRA SDEEKINPPP YLTCESFPHA VDHILQHLL | |
Greater than 95% as determined by SDS-PAGE. |
HDHD2 Protein | |
Research Use Only | |
HDHD2 | |
Human | |
His | |
The HDHD2 protein solution (1 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 1mM DTT, and 10% glycerol. |