missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GM-CSF / CSF2 Protein

A DNA sequence encoding the Human GM-CSF / CSF2, was expressed in the hosts and tags indicated.

Marke:  enQuireBio™ QP10346-ec-100ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15983568

  • 1120.00 CHF / 100 Mikrogramm


Verfügbar ab
30-10-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

NP_000749.2
GM-CSF / CSF2 Protein
100 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
APARSPSPST QPWEHVNAIQ EARRLLNLSR DTAAEMNETV EVISEMFDLQ EPTCLQTRLE LYKQGLRGSL TKLKGPLTMM ASHYKQHCPP TPETSCATQI ITFESFKENL KDFLLVIPFD CWEPVQE
1437
Greater than 98% as determined by SDS-PAGE and HPLC.
Research Use Only
CSF2
The ED50 as determined by a cell proliferation assay using human TF-1 cells is less than 0.1 ng/ml, corresponding to a specific activity of > 1.0 M 107 IU/mg.
Human
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
Dokumentation