missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GDNF Protein
A DNA sequence encoding the Human GDNF, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10417-ec-100ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_000505.1 | |
GDNF Protein | |
Research Use Only | |
GDNF | |
The ED50 as determined by a cell proliferation assay using rat C6 cells is less than 0.1 ng/ml, corresponding to a specific activity of > 5.0 M 107 IU/mg. | |
Human | |
Untagged | |
0.2 m filtered 1 M PBS, pH 7.4, with 0.05 % Tween-20, lyophilized |
2668 | |
100 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
SPDKQMAVLP RRERNRQAAA ANPENSRGKG RRGQRGKNRG CVLTAIHLNV TDLGLGYETK EELIFRYCSG SCDAAETTYD KILKNLSRNR RLVSDKVGQA CCRPIAFDDD LSFLDDNLVY HILRKHSAKR CGCI | |
Greater than 97% as determined by SDS-PAGE and HPLC. |