missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GDNF Protein

A DNA sequence encoding the Human GDNF, was expressed in the hosts and tags indicated.

Marke:  enQuireBio™ QP10417-ec-2ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15983788

  • 123.00 CHF / 2 Mikrogramm


Verfügbar ab
29-10-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

NP_000505.1
GDNF Protein
2 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
SPDKQMAVLP RRERNRQAAA ANPENSRGKG RRGQRGKNRG CVLTAIHLNV TDLGLGYETK EELIFRYCSG SCDAAETTYD KILKNLSRNR RLVSDKVGQA CCRPIAFDDD LSFLDDNLVY HILRKHSAKR CGCI
2668
Greater than 97% as determined by SDS-PAGE and HPLC.
Research Use Only
GDNF
The ED50 as determined by a cell proliferation assay using rat C6 cells is less than 0.1 ng/ml, corresponding to a specific activity of > 5.0 M 107 IU/mg.
Human
Untagged
0.2 m filtered 1 M PBS, pH 7.4, with 0.05 % Tween-20, lyophilized
Videos
Dokumentation