missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human G3BP2 Protein
A cDNA sequence encoding the G3BP2 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP11916-5ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
G3BP2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMVMEKP SPLLVGREFV RQYYTLLNKA PEYLHRFYGR NSSYVHGGVD ASGKPQEAVY GQNDIHHKVL SLNFSECHTK IRHVDAHATL SDGVVVQVMG LLSNSGQPER KFMQTFVLAP EGSVPNKFYV HNDMFRYEDE VFGDSEPELD EESEDEVEEE QEERQPSPEP VQENANSGYY EAHPVTNGIE EPLEESSHEP EPEPESETKT EELKPQVEEK NLEELEEKST TPPPAEPVSL PQEPPKPRVE AKPEVQSQPP RVREQRPRER PGFPPRGPRP GRGDMEQNDS DNRRIIRYPD SHQLFVGNLP HDIDENELKE FFMSFGNVVE LRINTKGVGG KLPNFGFVVF DDSEPVQRIL IAKPIMFRGE VRLNVEEKKT RAARERETRG GGDDRRDIRR NDRGPGGPRG IVGGGMMRDR DGRGPPPRGG MAQKLGSGRG TGQMEGRFTG QRR | |
Greater than 90% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The G3BP2 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 30% glycerol, 0.2M NaCl, 2mM DTT and 0.1mM PMSF. |