missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Fractalkine / CX3CL1 Protein

A DNA sequence encoding the Human Fractalkine / CX3CL1, was expressed in the hosts and tags indicated.

Marke:  enQuireBio™ QP10222-ec-250ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15983128

  • 2310.00 CHF / 250 Mikrogramm


Verfügbar ab
29-10-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

NP_001291321.1
Fractalkine / CX3CL1 Protein
250 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
QHHGVTKCNI TCSKMTSKIP VALLIHYQQN QASCGKRAII LETRQHRLFC ADPKEQWVKD AMQHLDRQAA ALTRNG
6376
Greater than 97% as determined by SDS-PAGE and HPLC.
Research Use Only
CX3CL1
The biological activity determined by a chemotaxis bioassay using human T-lymphocytes is in a concentration of 5.0-10 ng/ml.
Human
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
Dokumentation